1-Chlor-4-fluor-2-nitrobenzol

Lieferant: FLUOROCHEM

004456-5G 004456-25G
FLUO004456-5GEA 0 EUR
FLUO004456-5G FLUO004456-25G
1-Chlor-4-fluor-2-nitrobenzol
1-Chlor-4-fluor-2-nitrobenzol
Formel: C₆H₃ClFNO₂
Molecular Weight: 175,55 g/mol
Siedepunkt: 238,5 °C (1013 hPa)
Schmelzpunkt: 37…40 °C
MDL: MFCD00084853
CAS-Nummer: 345-17-5
EINECS: 206-456-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR