4-Chlor-3-fluorphenol

Lieferant: FLUOROCHEM

004459-5G 004459-25G 004459-10G
FLUO004459-5GEA 31.1 EUR
FLUO004459-5G FLUO004459-25G FLUO004459-10G
4-Chlor-3-fluorphenol
4-Chlor-3-fluorphenol
Formel: C₆H₄ClFO
Molecular Weight: 146,55 g/mol
Siedepunkt: 163 °C (1013 hPa)
Schmelzpunkt: 49…53 °C
MDL: MFCD00042583
CAS-Nummer: 348-60-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR