3-Chlor-4-fluorphenol
Lieferant: Thermo Scientific
SB01097.1
SB01097.10
SB01097.50
MAYBSB01097.1EA
0
EUR
MAYBSB01097.1
MAYBSB01097.10
MAYBSB01097.50
3-Chlor-4-fluorphenol
3-Chlor-4-fluorphenol
Formel:
C₆H₄ClFO Molecular Weight: 146,55 g/mol Siedepunkt: 225 °C (1013 hPa) Schmelzpunkt: 42…44 °C |
MDL:
MFCD00002257 CAS-Nummer: 2613-23-2 EINECS: 220-042-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...