1,2-Dichlor-4-iodbenzol
Lieferant: Thermo Scientific
Synonyme:
3,4-Dichloroiodobenzene
BTB06155.1
BTB06155EA
BTB06155.50
MAYBBTB06155.1EA
0
EUR
MAYBBTB06155.1
MAYBBTB06155EA
MAYBBTB06155.50
1,2-Dichlor-4-iodbenzol
1,2-Dichlor-4-iodbenzol
Formel:
C₆H₃Cl₂I Molecular Weight: 272,9 g/mol |
MDL:
MFCD00019014 CAS-Nummer: 20555-91-3 EINECS: 243-874-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...