1,2-Dichlor-4-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dichloroiodobenzene

001166-25G 001166-100G
FLUO001166-25GEA 60.9 EUR
FLUO001166-25G FLUO001166-100G
1,2-Dichlor-4-iodbenzol
1,2-Dichlor-4-iodbenzol
Formel: C₆H₃Cl₂I
MDL: MFCD00019014
CAS-Nummer: 20555-91-3
EINECS: 243-874-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR