1,3-Dichlor-5-iodbenzol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dichloroiodobenzene

A13192.06 A13192.14 A13192.22
A13192.06EA 40.8 EUR
A13192.06 A13192.14 A13192.22
1,3-Dichlor-5-iodbenzol ≥99%
1,3-Dichlor-5-iodbenzol

Purity: 99% CAS No.: 3032-81-3 Molecular Formula: C6H3Cl2I Formula Weight: 272.90 Melting Point: 55-59° Boiling Point: 127°/20mm MDL No.: MFCD00001047

Formel: C₆H₃Cl₂I
Molecular Weight: 272,9 g/mol
Siedepunkt: 127 °C (20 mmHg)
Schmelzpunkt: 55…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001047
CAS-Nummer: 3032-81-3
EINECS: 221-216-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR