4-Chlor-2-iodanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25457.14
B25457.06
B25457.14EA
352
EUR
B25457.14
B25457.06
4-Chlor-2-iodanilin ≥98%
4-Chlor-2-iodanilin
Purity: 98%
CAS No.: 63069-48-7
Molecular Formula: C6H5ClIN
Formula Weight: 253.47
Flash Point:
Formel:
C₆H₅ClIN Schmelzpunkt: 41…44 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863737 CAS-Nummer: 63069-48-7 EINECS: 263-839-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...