4-Chlor-2-iodanilin

Lieferant: FLUOROCHEM

005435-25G 005435-100G
FLUO005435-25GEA 63.9 EUR
FLUO005435-25G FLUO005435-100G
4-Chlor-2-iodanilin
4-Chlor-2-iodanilin
Formel: C₆H₅ClIN
MDL: MFCD01863737
CAS-Nummer: 63069-48-7
EINECS: 263-839-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR