2,4,6-Trichloranilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A19044.14
A19044.22
A19044.14EA
31.7
EUR
A19044.14
A19044.22
2,4,6-Trichloranilin ≥98%
2,4,6-Trichloranilin
Purity: 98+%
CAS No.: 634-93-5
Molecular Formula: C6H4Cl3N
Formula Weight: 196.46
Melting Point: 75-79°
Boiling Point: 262°
UN No.: 2018
MDL No.: MFCD00007663
Formel:
C₆H₄Cl₃N Molecular Weight: 196.46 g/mol Siedepunkt: 262 °C Schmelzpunkt: 75…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007663 CAS-Nummer: 634-93-5 EINECS: 211-219-8 UN: 2018 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...