3,4-Dichloranilin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
113110010
113110050
113112500
ACRO113110010EA
182
EUR
ACRO113110010
ACRO113110050
ACRO113112500
3,4-Dichloranilin 98%
3,4-Dichloranilin
Formel:
C₆H₅Cl₂N Molecular Weight: 162.02 g/mol Siedepunkt: 272 Schmelzpunkt: 69…73 °C Flammpunkt: 166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007768 CAS-Nummer: 95-76-1 EINECS: 202-448-4 UN: 3442 ADR: 6.1 II Merck Index: 13,03079 |
Spezifikation Testergebnisse
Appearance | White to grey to brown Crystals or flakes or crystalline mass and/or chunks (May darken on storage) |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
GC | ≥97.5 % |
Specific optical rotation | On anhydrous basis |
Titration with HClO4 | 97.5 to 102.5 % (On anhydrous basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...