2-Chlor-4-nitroanilin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-chlornitrobenzol
A16586.0B
A16586.0I
A16586.30
A16586.0BEA
128
EUR
A16586.0B
A16586.0I
A16586.30
2-Chlor-4-nitroanilin ≥98%
2-Chlor-4-nitroanilin
Purity: 98+%
CAS No.: 121-87-9
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Flash Point: 193°(379°F)
Odour: Weak, characteristic
Boiling Point: 320° dec.
Beilstein Reference: 638657
UN No.: 2237
MDL No.: MFCD00007665
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172,57 g/mol Siedepunkt: 320 °C (Zers.) Schmelzpunkt: 106…109 °C Dichte: 1 g/cm³ (20 °C) Flammpunkt: 193 °C (379 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007665 CAS-Nummer: 121-87-9 EINECS: 204-502-2 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...