2-Chlor-5-nitroanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15102.22
A15102.36
A15102.22EA
27.5
EUR
A15102.22
A15102.36
2-Chlor-5-nitroanilin ≥98%
2-Chlor-5-nitroanilin
Purity: 98%
CAS No.: 6283-25-6
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Flash Point: 191°(375°F)
Odour: Weak, characteristic
Beilstein Reference: 2208878
UN No.: 2237
MDL No.: MFCD00007668
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172.57 g/mol Schmelzpunkt: 117…121 °C Flammpunkt: 191 °C (375 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007668 CAS-Nummer: 6283-25-6 EINECS: 228-498-7 UN: 2237 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...