4-Chlor-2-nitroanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
PCONA
A18143.0B
A18143.0I
A18143.30
A18143.0BEA
424
EUR
A18143.0B
A18143.0I
A18143.30
4-Chlor-2-nitroanilin ≥98%
4-Chlor-2-nitroanilin
Purity: 98%
CAS No.: 89-63-4
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Flash Point: 191°(375°F)
Odour: Weak, characteristic
Beilstein Reference: 512436
UN No.: 2237
MDL No.: MFCD00007836
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172.57 g/mol Schmelzpunkt: 114…118 °C Dichte: 1.37 Flammpunkt: 191 °C (375 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007836 CAS-Nummer: 89-63-4 EINECS: 201-925-4 UN: 2237 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...