1,4-Dichlor-2-nitrobenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Nitro-2,5-dichlorbenzol, 2,5-Dichlor-1-nitrobenzol
A11896.0E
A11896.22
A11896.36
A11896.0EEA
195
EUR
A11896.0E
A11896.22
A11896.36
1,4-Dichlor-2-nitrobenzol ≥98%
1,4-Dichlor-2-nitrobenzol
Purity: 98%
CAS No.: 89-61-2
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Flash Point: 125°(257°F)
Melting Point: 53-57°
Boiling Point: 266-269°
Beilstein Reference: 778109
UN No.: 3077
MDL No.: MFCD00007074
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Siedepunkt: 266…269 °C Schmelzpunkt: 53…57 °C Dichte: 1,6 Flammpunkt: 125 °C (257 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007074 CAS-Nummer: 89-61-2 EINECS: 201-923-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...