1,3,5-Trichlor-2-nitrobenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Nitro-2,4,6-trichlorbenzol, 2,4,6-Trichlornitrobenzol
A12262.06
A12262.14
A12262.22
A12262.06EA
22.8
EUR
A12262.06
A12262.14
A12262.22
1,3,5-Trichlor-2-nitrobenzol ≥98%
1,3,5-Trichlor-2-nitrobenzol
Purity: 98+%
CAS No.: 18708-70-8
Molecular Formula: C6H2Cl3NO2
Formula Weight: 226.45
Melting Point: 68-73°
Beilstein Reference: 2213282
UN No.: 2811
MDL No.: MFCD00014690
Formel:
C₆H₂Cl₃NO₂ Molecular Weight: 226.45 g/mol Schmelzpunkt: 68…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014690 CAS-Nummer: 18708-70-8 EINECS: 242-518-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...