2-Chlor-5-nitroanisol
Lieferant: FLUOROCHEM
Synonyme:
1-Chloro-2-methoxy-4-nitrobenzene
003636-25G
003636-100G
FLUO003636-25GEA
104
EUR
FLUO003636-25G
FLUO003636-100G
2-Chlor-5-nitroanisol
2-Chlor-5-nitroanisol
Formel:
C₇H₆ClNO₃ Molecular Weight: 187.58 g/mol |
MDL:
MFCD00079739 CAS-Nummer: 1009-36-5 EINECS: 213-768-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...