4-Chlor-3-nitrophenol
Lieferant: CHEMPUR
|
Achtung
|
FL13480.POR
013480-25G
013480-5G
CHMPFL13480.POREA
0
EUR
CHMPFL13480.POR
FLUO013480-25G
FLUO013480-5G
4-Chlor-3-nitrophenol
4-Chlor-3-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173,56 g/mol Schmelzpunkt: 125…128 °C Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043546 CAS-Nummer: 610-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...