2,5-Dichlorphenol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,5-DCP

113580500 113582500 113580010
ACRO113580500EA 32 EUR
ACRO113580500 ACRO113582500 ACRO113580010
2,5-Dichlorphenol 98%
2,5-Dichlorphenol
Formel: C₆H₄Cl₂O
Molecular Weight: 163 g/mol
Siedepunkt: 211 °C (760 mmHg)
Schmelzpunkt: 54…58 °C
Flammpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002174
CAS-Nummer: 583-78-8
EINECS: 209-520-4
UN: 2020
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystals of flakes
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %
Uniformity coefficient ≤1.60
Moisture holding capacity 49.0 to 60.0 % (Cl)
Total volume capacity ≥1.20 eq/l (Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR