2,3-Dichlorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 2,3-Dichlorphenylmercaptan, 1,2-Dichlor-3-mercaptobenzol, 2,3-Dichlorbenzol-1-thiol, 2,3-Dichlorbenzolthiol

003351-5G 003351-25G
FLUO003351-5GEA 64.7 EUR
FLUO003351-5G FLUO003351-25G
2,3-Dichlorthiophenol
2,3-Dichlorthiophenol
Formel: C₆H₄Cl₂S
Molecular Weight: 179.07 g/mol
Siedepunkt: 116…118 °C (20 mmHg)
Schmelzpunkt: 46.5
MDL: MFCD00041420
CAS-Nummer: 17231-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR