2-Chlorthiophenol

Lieferant: FLUOROCHEM

Synonyme: o-Chlorthiophenol, 2-Chlorbenzol-1-thiol, 1-Chlor-2-mercaptobenzol, 2-Chlorbenzolthiol, 2-Chlorphenylmercaptan

001580-25G 001580-100G
FLUO001580-25GEA 228 EUR
FLUO001580-25G FLUO001580-100G
2-Chlorthiophenol
2-Chlorthiophenol
Formel: C₆H₅ClS
Molecular Weight: 144.62 g/mol
MDL: MFCD00004830
CAS-Nummer: 6320-03-2
EINECS: 228-667-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR