3-Chlorthiophenol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Chlor-3-mercaptobenzol, 3-Chlorphenylmercaptan, 3-Chlorbenzol-1-thiol, m-Chlorthiophenol, 3-Chlorbenzolthiol
L04806.06
L04806.14
L04806.03
L04806.06EA
51.2
EUR
L04806.06
L04806.14
L04806.03
3-Chlorthiophenol ≥97%
3-Chlorthiophenol
Purity: 97%
CAS No.: 2037-31-2
Molecular Formula: C6H5ClS
Formula Weight: 144.62
Flash Point: 90°(194°F)
Refractive Index: 1.6021
Boiling Point: 104-105°/35mm
Beilstein Reference: 1854087
UN No.: 1760
MDL No.: MFCD00004839
Formel:
C₆H₅ClS Molecular Weight: 144.62 g/mol Siedepunkt: 104…105 °C (35 mmHg) Dichte: 1.244 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004839 CAS-Nummer: 2037-31-2 EINECS: 218-010-0 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...