3-Chlorthiophenol
Lieferant: Thermo Scientific
Synonyme:
1-Chlor-3-mercaptobenzol, 3-Chlorphenylmercaptan, 3-Chlorbenzol-1-thiol, m-Chlorthiophenol, 3-Chlorbenzolthiol
BTB06490.1
BTB06490.10
BTB06490.50
MAYBBTB06490.1EA
0
EUR
MAYBBTB06490.1
MAYBBTB06490.10
MAYBBTB06490.50
3-Chlorthiophenol
3-Chlorthiophenol
Formel:
C₆H₅ClS Molecular Weight: 144.62 g/mol Siedepunkt: 110 Dichte: 1.2637 g/cm³ (12.5 °C) |
MDL:
MFCD00004839 CAS-Nummer: 2037-31-2 EINECS: 218-010-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...