3-Chlorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-3-mercaptobenzol, 3-Chlorphenylmercaptan, 3-Chlorbenzol-1-thiol, m-Chlorthiophenol, 3-Chlorbenzolthiol

001581-5G 001581-25G 001581-100G
FLUO001581-5GEA 87.9 EUR
FLUO001581-5G FLUO001581-25G FLUO001581-100G
3-Chlorthiophenol
3-Chlorthiophenol
Formel: C₆H₅ClS
Molecular Weight: 144.62 g/mol
Siedepunkt: 110
Dichte: 1.2637 g/cm³ (12.5 °C)
MDL: MFCD00004839
CAS-Nummer: 2037-31-2
EINECS: 218-010-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR