1,2,3-Trifluorbenzol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
B22055.03
B22055.06
B22055.03EA
24.5
EUR
B22055.03
B22055.06
1,2,3-Trifluorbenzol ≥98%
1,2,3-Trifluorobenzene
Purity: 98+%
CAS No.: 1489-53-8
Molecular Formula: C6H3F3
Formula Weight: 132.09
Flash Point: -3°(26°F)
Refractive Index: 1.4220
Boiling Point: 94-95°
Beilstein Reference: 1862388
UN No.: 2387
MDL No.: MFCD00012013
Formel:
C₆H₃F₃ Molecular Weight: 132,09 g/mol Siedepunkt: 94…95 °C Dichte: 1,280 Flammpunkt: –3 °C (26 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012013 CAS-Nummer: 1489-53-8 UN: 2387 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...