1,3,5-Trifluorbenzol
Lieferant: FLUOROCHEM
001585-25G
001585-100G
001585-10G
FLUO001585-25GEA
55.4
EUR
FLUO001585-25G
FLUO001585-100G
FLUO001585-10G
1,3,5-Trifluorbenzol
1,3,5-Trifluorobenzene
Formel:
C₆H₃F₃ Molecular Weight: 132.09 g/mol Siedepunkt: 75 Dichte: 1.277 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000333 CAS-Nummer: 372-38-3 EINECS: 206-751-2 UN: 2387 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...