Hexafluorbenzol ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Perfluorobenzene

A11500.PF A11500.0I
A11500.PFEA 0 EUR
A11500.PF A11500.0I
Hexafluorbenzol ≥99%
Hexafluorbenzol

Purity: 99% CAS No.: 392-56-3 Molecular Formula: C6F6 Formula Weight: 186.06 Flash Point: 10°(50°F) Refractive Index: 1.3780 Odour: Aromatic Melting Point: 4-5° Boiling Point: 80-81° Beilstein Reference: 1683438 Merck Reference: 14,4686 UN No.: 1993 MDL No.: MFCD00000288

Formel: C₆F₆
Molecular Weight: 186,06 g/mol
Siedepunkt: 80…81 °C
Schmelzpunkt: 4…5 °C
Dichte: 1,616
Flammpunkt: 10 °C (50 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000288
CAS-Nummer: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,II
Merck Index: 14,4686

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR