2,3,5,6-Tetrafluorbenzaldehyd
Lieferant: FLUOROCHEM
|
Achtung
|
009870-1G
009870-5G
FLUO009870-1GEA
31.1
EUR
FLUO009870-1G
FLUO009870-5G
2,3,5,6-Tetrafluorbenzaldehyd
2,3,5,6-Tetrafluorobenzaldehyde
Formel:
C₇H₂F₄O Molecular Weight: 178.09 g/mol |
MDL:
MFCD00167387 CAS-Nummer: 19842-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...