3,4-Difluorbenzaldehyd
Lieferant: FLUOROCHEM
001464-25G
001464-100G
001464-10G
FLUO001464-25GEA
60.9
EUR
FLUO001464-25G
FLUO001464-100G
FLUO001464-10G
3,4-Difluorbenzaldehyd
3,4-Difluorobenzaldehyde
Formel:
C₇H₄F₂O Molecular Weight: 142,1 g/mol Siedepunkt: 53 °C (1013 hPa) Dichte: 1,29 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010328 CAS-Nummer: 34036-07-2 EINECS: 422-180-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...