2,3-Difluorbenzonitril

Lieferant: FLUOROCHEM

001859-5G 001859-25G
FLUO001859-5GEA 36.9 EUR
FLUO001859-5G FLUO001859-25G
2,3-Difluorbenzonitril
2,3-Difluorbenzonitril
Formel: C₇H₃F₂N
Molecular Weight: 139,1 g/mol
Siedepunkt: 189 °C (1013 hPa)
Dichte: 1,25 g/cm³ (20 °C)
MDL: MFCD00009976
CAS-Nummer: 21524-39-0
EINECS: 244-418-3
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR