2,4,5-Trifluorbenzonitril

Lieferant: FLUOROCHEM

001874-5G 001874-25G
FLUO001874-5GEA 0 EUR
FLUO001874-5G FLUO001874-25G
2,4,5-Trifluorbenzonitril
2,4,5-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157,09 g/mol
Siedepunkt: 168…170 °C (1013 hPa)
Dichte: 1,37 g/cm³ (20 °C)
MDL: MFCD00013289
CAS-Nummer: 98349-22-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR