2,4,6-Trifluorbenzonitril

Lieferant: FLUOROCHEM

007487-5G 007487-25G
FLUO007487-5GEA 39.9 EUR
FLUO007487-5G FLUO007487-25G
2,4,6-Trifluorbenzonitril
2,4,6-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157.09 g/mol
Siedepunkt: 179
Schmelzpunkt: 57…58 °C
MDL: MFCD00042399
CAS-Nummer: 96606-37-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR