2,4-Difluorbenzonitril
Lieferant: FLUOROCHEM
001918-25G
001918-100G
001918-500G
FLUO001918-25GEA
34.1
EUR
FLUO001918-25G
FLUO001918-100G
FLUO001918-500G
2,4-Difluorbenzonitril
2,4-Difluorbenzonitril
Formel:
C₇H₃F₂N Molecular Weight: 139.1 g/mol Siedepunkt: 80…83 °C (22.5 mmHg) Schmelzpunkt: 46…47 °C |
MDL:
MFCD00009826 CAS-Nummer: 3939-09-1 EINECS: 223-523-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...