3,4-Difluorbenzonitril

Lieferant: FLUOROCHEM

002081-25G 002081-100G
FLUO002081-25GEA 35.3 EUR
FLUO002081-25G FLUO002081-100G
3,4-Difluorbenzonitril
3,4-Difluorbenzonitril
Formel: C₇H₃F₂N
Molecular Weight: 139.1 g/mol
Siedepunkt: 189
Schmelzpunkt: 51
MDL: MFCD00011666
CAS-Nummer: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR