3,5-Difluorbenzonitril

Lieferant: FLUOROCHEM

002111-10G 002111-250G 002111-50G
FLUO002111-10GEA 0 EUR
FLUO002111-10G FLUO002111-250G FLUO002111-50G
3,5-Difluorbenzonitril
3,5-Difluorbenzonitril
Formel: C₇H₃F₂N
Siedepunkt: 159…160 °C (1013 hPa)
MDL: MFCD00010311
CAS-Nummer: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR