2,3,4-Trifluorbenzoesäure

Lieferant: Thermo Scientific
Achtung

SB01766.1 SB01766.10
MAYBSB01766.1EA 0 EUR
MAYBSB01766.1 MAYBSB01766.10
2,3,4-Trifluorbenzoesäure
2,3,4-Trifluorobenzoic acid
Formel: C₇H₃F₃O₂
Molecular Weight: 176.09 g/mol
Siedepunkt: 237
Schmelzpunkt: 140…142 °C
MDL: MFCD00061232
CAS-Nummer: 61079-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR