2,3,4-Trifluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

001808-25G 001808-100G 001808-250G
FLUO001808-25GEA 36.9 EUR
FLUO001808-25G FLUO001808-100G FLUO001808-250G
2,3,4-Trifluorbenzoesäure
2,3,4-Trifluorobenzoic acid
Formel: C₇H₃F₃O₂
Molecular Weight: 176.09 g/mol
Siedepunkt: 237
Schmelzpunkt: 140…142 °C
MDL: MFCD00061232
CAS-Nummer: 61079-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR