2,3-Difluorbenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
CFF613.5
001858-5G
001858-25G
001858-100G
CHMPCFF613.5EA
401
EUR
CHMPCFF613.5
FLUO001858-5G
FLUO001858-25G
FLUO001858-100G
2,3-Difluorbenzoesäure
2,3-Difluorobenzoic acid
Formel:
C₇H₄F₂O₂ Molecular Weight: 158,1 g/mol |
MDL:
MFCD00010267 CAS-Nummer: 4519-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...