2,4,5-Trifluorbenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

A14193.06 A14193.14
A14193.06EA 54.6 EUR
A14193.06 A14193.14
2,4,5-Trifluorbenzoesäure ≥98%
2,4,5-Trifluorobenzoic acid

Purity: 98% CAS No.: 446-17-3 Molecular Formula: C7H3F3O2 Formula Weight: 176.10 Melting Point: 95-100° Beilstein Reference: 3257609

Formel: C₇H₃F₃O₂
Molecular Weight: 176,09 g/mol
Siedepunkt: 237 °C (1013 hPa)
Schmelzpunkt: 95…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013306
CAS-Nummer: 446-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR