2,4,5-Trifluorbenzoesäure
Lieferant: Thermo Scientific
|
Achtung
|
SB01325.1
SB01325.10
MAYBSB01325.1EA
0
EUR
MAYBSB01325.1
MAYBSB01325.10
2,4,5-Trifluorbenzoesäure
2,4,5-Trifluorobenzoic acid
Formel:
C₇H₃F₃O₂ Molecular Weight: 176.09 g/mol Siedepunkt: 237 Schmelzpunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013306 CAS-Nummer: 446-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...