4-Fluoranthranilsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-4-fluorobenzoic acid

B21907.03 B21907.06
B21907.03EA 63.4 EUR
B21907.03 B21907.06
4-Fluoranthranilsäure 98%
4-Fluoranthranilsäure

Purity: 98% CAS No.: 446-32-2 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 192-196° Beilstein Reference: 2803665

Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
Siedepunkt: 305 °C (1013 hPa)
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075553
CAS-Nummer: 446-32-2
EINECS: 207-163-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR