5-Fluoranthranilsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-fluorobenzoic acid
B21458.03
B21458.06
B21458.14
B21458.03EA
61.5
EUR
B21458.03
B21458.06
B21458.14
5-Fluoranthranilsäure ≥98%
5-Fluoranthranilsäure
Purity: 98+%
CAS No.: 446-08-2
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Melting Point: 182-184°
Beilstein Reference: 2803664
Formel:
C₇H₆FNO₂ Molecular Weight: 155,13 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 182…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055566 CAS-Nummer: 446-08-2 EINECS: 207-159-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...