4-Amino-2,3,5,6-tetrafluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

003133-1G 003133-5G
FLUO003133-1GEA 51.8 EUR
FLUO003133-1G FLUO003133-5G
4-Amino-2,3,5,6-tetrafluorbenzoesäure
4-Amino-2,3,5,6-tetrafluorbenzoesäure
Formel: C₇H₃F₄NO₂
Molecular Weight: 209.1 g/mol
MDL: MFCD00007647
CAS-Nummer: 944-43-4
EINECS: 213-409-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR