4-Amino-3-fluorbenzoesäure

Lieferant: BIONET RESEARCH
Achtung

Synonyme: 4-Amino-3-fluorobenzoic Acid

3T-0232.500 3T-0232.1
BIOT3T-0232.500EA 0 EUR
BIOT3T-0232.500 BIOT3T-0232.1
4-Amino-3-fluorbenzoesäure
4-Amino-3-fluorobenzoic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
MDL: MFCD01660374
CAS-Nummer: 455-87-8
EINECS: 207-250-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR