4-Amino-3-fluorbenzoesäure

Lieferant: CHEMPUR
Achtung

Synonyme: 4-Amino-3-fluorobenzoic Acid

CFF775. FL13032.POR 013032-5G 013032-1G 013032-25G
CHMPCFF775.EA 0 EUR
CHMPCFF775. CHMPFL13032.POR FLUO013032-5G FLUO013032-1G FLUO013032-25G
4-Amino-3-fluorbenzoesäure
4-Amino-3-fluorobenzoic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
MDL: MFCD01660374
CAS-Nummer: 455-87-8
EINECS: 207-250-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR