2-Fluor-4-nitrobenzoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L19770.03
L19770.06
L19770.03EA
73.5
EUR
L19770.03
L19770.06
2-Fluor-4-nitrobenzoesäure ≥98%
2-Fluor-4-nitrobenzoesäure
Purity: 98%
CAS No.: 403-24-7
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 172-175°
Beilstein Reference: 2582091
Formel:
C₇H₄FNO₄ Molecular Weight: 185,11 g/mol Schmelzpunkt: 172…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00275565 CAS-Nummer: 403-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...