2-Fluor-4-nitrobenzoesäure

Lieferant: BIONET RESEARCH
Achtung

7R-0042.1 7R-0042.10 7R-0042.5 7R-0042.500
BIOT7R-0042.1EA 0 EUR
BIOT7R-0042.1 BIOT7R-0042.10 BIOT7R-0042.5 BIOT7R-0042.500
2-Fluor-4-nitrobenzoesäure
2-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
MDL: MFCD00275565
CAS-Nummer: 403-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR