2-Fluor-4-nitrobenzoesäure
Lieferant: Thermo Scientific
|
Achtung
|
TL00808DA
TL00808EA
MAYBTL00808DAEA
0
EUR
MAYBTL00808DA
MAYBTL00808EA
2-Fluor-4-nitrobenzoesäure
2-Fluor-4-nitrobenzoesäure
Formel:
C₇H₄FNO₄ Molecular Weight: 185,11 g/mol |
MDL:
MFCD00275565 CAS-Nummer: 403-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...