2-Fluor-4-nitrobenzoesäure

Lieferant: FLUOROCHEM
Achtung

008351-1G 008351-5G
FLUO008351-1GEA 31.1 EUR
FLUO008351-1G FLUO008351-5G
2-Fluor-4-nitrobenzoesäure
2-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00275565
CAS-Nummer: 403-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR