3-(Trifluormethoxy)benzoesäure
Lieferant: CHEMPUR
|
Achtung
|
SB01718.250
SB01718.1
CHMPSB01718.250EA
0
EUR
CHMPSB01718.250
MAYBSB01718.1
3-(Trifluormethoxy)benzoesäure
3-(Trifluormethoxy)benzoesäure
Formel:
C₈H₅F₃O₃ Molecular Weight: 206.12 g/mol Siedepunkt: 272 Schmelzpunkt: 91…92 °C |
MDL:
MFCD00041501 CAS-Nummer: 1014-81-9 EINECS: 213-802-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...