1,2-Diiodtetrafluorbenzol

Lieferant: FLUOROCHEM
Achtung

010455-5G 010455-1G
FLUO010455-5GEA 0 EUR
FLUO010455-5G FLUO010455-1G
1,2-Diiodtetrafluorbenzol
1,2-Diiodtetrafluorbenzol
Formel: C₆F₄I₂
MDL: MFCD00019011
CAS-Nummer: 2708-97-6
EINECS: 220-303-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR